accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClBr

CAS RN 109-70-6

IUPAC Name: 1-bromo-3-chloropropane
Synonyms: 1-chloro-3-bromopropane
Molecular Weight (g/mol): 157.44
Molecular Formula: C3H6BrCl
InChi Key: MFESCIUQSIBMSM-UHFFFAOYSA-N
SMILES: ClCCCBr
Boiling Point 
  • (4)
  • (4)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (6)
Percent Purity 
  • (2)
  • (10)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (8)
  • (4)
  • (4)
  • (2)
  • (2)
  • (6)
  • (6)
  • (2)
  • (10)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)