accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109-81-9

IUPAC Name: (2-azaniumylethyl)(methyl)azanium
Synonyms: (2-ammonioethyl)(methyl)azanium
Molecular Weight (g/mol): 76.14
Molecular Formula: C3H12N2
InChi Key: KFIGICHILYTCJF-UHFFFAOYSA-P
SMILES: C[NH2+]CC[NH3+]
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (9)
Percent Purity 
  • (3)
  • (9)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (9)
  • (3)
  • (9)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (6)