accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1091606-67-5

IUPAC Name: [(1S,2S)-2-amino-1,2-diphenylethyl]diphenylphosphane
Synonyms: [(1S,2S)-2-amino-1,2-diphenylethyl]diphenylphosphane
Molecular Weight (g/mol): 381.46
Molecular Formula: C26H24NP
InChi Key: MIPGTOGPLXZBQX-QYRYXDJWNA-N
SMILES: N[C@H]([C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results