accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1091606-70-0

IUPAC Name: 2-(diphenylphosphanyl)-2,3-dihydro-1H-inden-1-amine
Synonyms: 2-(diphenylphosphanyl)-2,3-dihydro-1H-inden-1-amine
Molecular Weight (g/mol): 317.37
Molecular Formula: C21H20NP
InChi Key: IALYUOSURKZWLR-UHFFFAOYNA-N
SMILES: NC1C(CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results