accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1092349-87-5

IUPAC Name: 2-bromo-3,4-difluoroaniline
Synonyms: 2-bromo-3,4-difluoroaniline
Molecular Weight (g/mol): 208.01
Molecular Formula: C6H4BrF2N
InChi Key: ZGZURPPCPVPDMX-UHFFFAOYSA-N
SMILES: NC1=C(Br)C(F)=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results