accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONH2OH2N

CAS RN 109244-58-8

IUPAC Name: methyl 2-(3,6-diamino-9H-xanthen-9-yl)benzoate
Synonyms: methyl 2-(3,6-diamino-9H-xanthen-9-yl)benzoate
Molecular Weight (g/mol): 346.39
Molecular Formula: C21H18N2O3
InChi Key: FNEZBBILNYNQGC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1C1C2=CC=C(N)C=C2OC2=CC(N)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results