accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1092522-75-2

IUPAC Name: tridecacyclo[44.2.2.2²,⁵.2⁶,⁹.2¹⁰,¹³.2¹⁴,¹⁷.2¹⁸,²¹.2²²,²⁵.2²⁶,²⁹.2³⁰,³³.2³⁴,³⁷.2³⁸,⁴¹.2⁴²,⁴⁵]doheptaconta-1(49),2,4,6,8,10,12,14,16,18,20,22,24,26,28,30,32,34(56),35,37(55),38,40,42(52),43,45(51),46(50),47,53,57,59,61,63,65,67,69,71-hexatriacontaene
Synonyms: tridecacyclo[44.2.2.2²,⁵.2⁶,⁹.2¹⁰,¹³.2¹⁴,¹⁷.2¹⁸,²¹.2²²,²⁵.2²⁶,²⁹.2³⁰,³³.2³⁴,³⁷.2³⁸,⁴¹.2⁴²,⁴⁵]doheptaconta-1(49),2,4,6,8,10,12,14,16,18,20,22,24,26,28,30,32,34(56),35,37(55),38,40,42(52),43,45(51),46(50),47,53,57,59,61,63,65,67,69,71-hexatriacontaene
Molecular Weight (g/mol): 913.18
Molecular Formula: C72H48
InChi Key: AAFTYBVDGIFJMP-UHFFFAOYSA-N
SMILES: C1=CC2=CC=C1C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C2C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results