accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNHNNNH2N

CAS RN 1092788-83-4

IUPAC Name: 1-cyclopentyl-3-{1H-pyrrolo[2,3-b]pyridin-5-yl}-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Synonyms: 1-cyclopentyl-3-{1H-pyrrolo[2,3-b]pyridin-5-yl}pyrazolo[3,4-d]pyrimidin-4-amine
Molecular Weight (g/mol): 319.37
Molecular Formula: C17H17N7
InChi Key: NVRXTLZYXZNATH-UHFFFAOYSA-N
SMILES: NC1=C2C(=NC=N1)N(N=C2C1=CN=C2NC=CC2=C1)C1CCCC1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results