accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FNOOF

CAS RN 1093758-82-7

IUPAC Name: 2,5-difluoro-1-methyl-3-nitrobenzene
Synonyms: 2,5-difluoro-1-methyl-3-nitrobenzene
Molecular Weight (g/mol): 173.12
Molecular Formula: C7H5F2NO2
InChi Key: JPBHCSGGXITIHO-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC(=C1F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results