accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109384-19-2

IUPAC Name: tert-butyl 4-hydroxypiperidine-1-carboxylate
Synonyms: tert-butyl 4-hydroxypiperidine-1-carboxylate
Molecular Weight (g/mol): 201.27
Molecular Formula: C10H19NO3
InChi Key: PWQLFIKTGRINFF-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(O)CC1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)