accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNNNH2

CAS RN 1093860-47-9

IUPAC Name: 5-(aminomethyl)-N-phenylpyrimidin-2-amine
Synonyms: 5-(aminomethyl)-N-phenylpyrimidin-2-amine
Molecular Weight (g/mol): 200.25
Molecular Formula: C11H12N4
InChi Key: IGMKIKUXMJLMOP-UHFFFAOYSA-N
SMILES: NCC1=CN=C(NC2=CC=CC=C2)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results