accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1093860-48-0

IUPAC Name: methyl 6-chloropyridazine-4-carboxylate
Synonyms: methyl 6-chloropyridazine-4-carboxylate
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: QYQHGLKSLGUINZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=NN=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results