accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109425-55-0

IUPAC Name: 5-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms: 5-[(tert-butoxycarbonyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 454.52
Molecular Formula: C25H30N2O6
InChi Key: JOOIZTMAHNLNHE-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)