accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOClF

CAS RN 1094362-88-5

IUPAC Name: 4-(chloromethyl)-N-[(4-fluorophenyl)methyl]benzamide
Synonyms: 4-(chloromethyl)-N-[(4-fluorophenyl)methyl]benzamide
Molecular Weight (g/mol): 277.72
Molecular Formula: C15H13ClFNO
InChi Key: IWRWVLGYZZVJJR-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNC(=O)C2=CC=C(CCl)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results