accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109461-44-1

IUPAC Name: 2-(4-chlorophenyl)-2-methylmorpholine
Synonyms: 2-(4-chlorophenyl)-2-methylmorpholine
Molecular Weight (g/mol): 211.69
Molecular Formula: C11H14ClNO
InChi Key: IJDDASQRAPIORY-UHFFFAOYNA-N
SMILES: CC1(CNCCO1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results