accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(E/Z)

CAS RN 109463-48-1

IUPAC Name: ethyl 4-(2-phenylethenyl)benzoate
Synonyms: ethyl 4-(2-phenylethenyl)benzoate
Molecular Weight (g/mol): 252.31
Molecular Formula: C17H16O2
InChi Key: RBQGAPDZGPQIDL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=CC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results