accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3FFFFFF

CAS RN 1095-77-8

IUPAC Name: 1-[1,1,1,3,3,3-hexafluoro-2-(4-methylphenyl)propan-2-yl]-4-methylbenzene
Synonyms: 1-[1,1,1,3,3,3-hexafluoro-2-(4-methylphenyl)propan-2-yl]-4-methylbenzene
Molecular Weight (g/mol): 332.29
Molecular Formula: C17H14F6
InChi Key: OWEIAGSMFHSSES-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(C1=CC=C(C)C=C1)(C(F)(F)F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results