accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1095-78-9

IUPAC Name: 4-[2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]aniline
Synonyms: 4-[2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]aniline
Molecular Weight (g/mol): 334.27
Molecular Formula: C15H12F6N2
InChi Key: BEKFRNOZJSYWKZ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)(C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)