accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHOONNH(S)H3CH3C

CAS RN 1095952-22-9

IUPAC Name: (4S)-3-[2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Synonyms: (4S)-3-(2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}acetyl)-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol): 424.45
Molecular Formula: C23H24N2O6
InChi Key: XUBXHZJERGBCEL-IBGZPJMESA-N
SMILES: CC1(C)OC[C@H](N1C(=O)CNC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)