accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1096-80-6

IUPAC Name: triphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium bromide
Synonyms: triphenyl-1,2λ⁵,3,4-tetrazol-2-ylium bromide
Molecular Weight (g/mol): 379.26
Molecular Formula: C19H15BrN4
InChi Key: YZSUTKMFUHJPNB-UHFFFAOYSA-M
SMILES: [Br-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results