accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFCH3NH3CFCH3

CAS RN 109705-14-8

IUPAC Name: 1-fluoro-2,4,6-trimethylpyridin-1-ium; tetrafluoroboranuide
Synonyms: 1-fluoro-2,4,6-trimethylpyridin-1-ium tetrafluoroborate
Molecular Weight (g/mol): 226.99
Molecular Formula: C8H11BF5N
InChi Key: RRNLYYDDEUOXBB-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC1=CC(C)=[N+](F)C(C)=C1
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results