accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOP

CAS RN 1099-45-2

IUPAC Name: ethyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Synonyms: triphenylcarbethoxymethylenephosphorane
Molecular Weight (g/mol): 348.38
Molecular Formula: C22H21O2P
InChi Key: IIHPVYJPDKJYOU-UHFFFAOYSA-N
SMILES: CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (3)
  • (8)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)