accessibility menu, dialog, popup

UserName

Viewing profile as user:
S

CAS RN 110-02-1

IUPAC Name: thiophene
Synonyms: thiophene thiofurfuran thiacyclopentadiene thiaphene thio-furan thiofuram thiofuran furan, thio- thiofen huile H50
Molecular Weight (g/mol): 84.14
Molecular Formula: C4H4S
InChi Key: YTPLMLYBLZKORZ-UHFFFAOYSA-N
SMILES: S1C=CC=C1
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (3)
  • (1)
  • (2)
  • (8)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (10)
  • (1)
  • (5)
  • (1)