CAS RN 110-34-9
IUPAC Name:
2-methylpropyl hexadecanoate
Synonyms:
isobutyl palmitate
Molecular Weight (g/mol):
312.54
Molecular Formula:
C20H40O2
InChi Key:
OJIBJRXMHVZPLV-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCC(=O)OCC(C)C
1
–
1
of
1
results
1
Isobutyl Palmitate 97.0+%, TCI America™
CAS: 110-34-9 Molecular Formula: C20H40O2 Molecular Weight (g/mol): 312.538 MDL Number: MFCD00059289 InChI Key: OJIBJRXMHVZPLV-UHFFFAOYSA-N Synonym: Palmitic Acid Isobutyl Ester PubChem CID: 66967 IUPAC Name: 2-methylpropyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(C)C