CAS RN 110-36-1
IUPAC Name:
butyl tetradecanoate
Synonyms:
butyl myristate
Molecular Weight (g/mol):
284.48
Molecular Formula:
C18H36O2
InChi Key:
DHAZIUXMHRHVMP-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCC(=O)OCCCC