accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 110-38-3

IUPAC Name: ethyl decanoate
Synonyms: ethyl caprate
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: RGXWDWUGBIJHDO-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)OCC
Boiling Point 
  • (2)
  • (2)
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (2)
  • (7)
  • (2)
  • (2)
  • (4)
  • (3)
  • (6)
  • (5)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)