accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110-39-4

IUPAC Name: octyl butanoate
Synonyms: caprylyl butyrate
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: PWLNAUNEAKQYLH-UHFFFAOYSA-N
SMILES: CCCCCCCCOC(=O)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results