accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110-64-5

IUPAC Name: (2Z)-but-2-ene-1,4-diol
Synonyms: 2-butene-1,4-diol (cis)
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: ORTVZLZNOYNASJ-UPHRSURJSA-N
SMILES: OC\C=C/CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results