accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110-69-0

IUPAC Name: (Z)-N-butylidenehydroxylamine
Synonyms: (Z)-N-butylidenehydroxylamine
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: KGGVGTQEGGOZRN-PLNGDYQASA-N
SMILES: CCC\C=N/O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results