CAS RN 110-87-2
3,4-Dihydro-2H-pyran, 99%
CAS: 110-87-2 Molecular Formula: C5H8O Molecular Weight (g/mol): 84.12 MDL Number: MFCD00006558 InChI Key: BUDQDWGNQVEFAC-UHFFFAOYSA-N Synonym: dihydropyran,3,4-dihydropyran,2,3-dihydropyran,2h-3,4-dihydropyran,2,3-dihydro-4h-pyran,dihydro-2h-pyran,2h-pyran, dihydro,2h-pyran, 3,4-dihydro,5,6-dihydro-4h-pyran,dihydropyrane PubChem CID: 8080 IUPAC Name: 3,4-dihydro-2H-pyran SMILES: C1CC=COC1
3,4-Dihydro-2H-pyran, 99%
CAS: 110-87-2 Molecular Formula: C5H8O Molecular Weight (g/mol): 84.118 MDL Number: MFCD00006558 InChI Key: BUDQDWGNQVEFAC-UHFFFAOYSA-N Synonym: dihydropyran,3,4-dihydropyran,2,3-dihydropyran,2h-3,4-dihydropyran,2,3-dihydro-4h-pyran,dihydro-2h-pyran,2h-pyran, dihydro,2h-pyran, 3,4-dihydro,5,6-dihydro-4h-pyran,dihydropyrane PubChem CID: 8080 IUPAC Name: 3,4-dihydro-2H-pyran SMILES: C1CC=COC1
3,4-Dihydro-2H-pyran 97.0+%, TCI America™
CAS: 110-87-2 Molecular Formula: C5H8O Molecular Weight (g/mol): 84.118 MDL Number: MFCD00006558 InChI Key: BUDQDWGNQVEFAC-UHFFFAOYSA-N Synonym: dihydropyran,3,4-dihydropyran,2,3-dihydropyran,2h-3,4-dihydropyran,2,3-dihydro-4h-pyran,dihydro-2h-pyran,2h-pyran, dihydro,2h-pyran, 3,4-dihydro,5,6-dihydro-4h-pyran,dihydropyrane PubChem CID: 8080 IUPAC Name: 3,4-dihydro-2H-pyran SMILES: C1CC=COC1
Sigma Aldrich 3,4-Dihydro-2H-Pyran
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Diethyl Sulfate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
3,4-Dihydro-2H-Pyran, SPEX CertiPrep™
CAS: 110-87-2 Molecular Formula: C5H8O Molecular Weight (g/mol): 84.118 InChI Key: BUDQDWGNQVEFAC-UHFFFAOYSA-N PubChem CID: 8080 IUPAC Name: 3,4-dihydro-2H-pyran SMILES: C1CC=COC1