accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110-88-3

IUPAC Name: 1,3,5-trioxane
Synonyms: trioxane marvosan formagene triossimetilene s-trixane trioxymethylen trimer formaldehyde formaldehyde, trimer metaformaldehyde triformol
Molecular Weight (g/mol): 90.08
Molecular Formula: C3H6O3
InChi Key: BGJSXRVXTHVRSN-UHFFFAOYSA-N
SMILES: C1OCOCO1
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (3)
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (4)
  • (2)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (4)
  • (2)
  • (7)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)
  • (4)
  • (2)