accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3O

CAS RN 110-93-0

IUPAC Name: 6-methylhept-5-en-2-one
Synonyms: 6-methyl-5-hepten-2-one sulcatone methylheptenone methylheptenone isomers
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: UHEPJGULSIKKTP-UHFFFAOYSA-N
SMILES: CC(C)=CCCC(C)=O
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)