accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110046-60-1

IUPAC Name: 5'-bromo-[2,2'-bithiophene]-5-carbaldehyde
Synonyms: 5'-bromo-[2,2'-bithiophene]-5-carbaldehyde
Molecular Weight (g/mol): 273.16
Molecular Formula: C9H5BrOS2
InChi Key: NTHMTYNJFSUBMF-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=CC=C(S1)C=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)