accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110223-15-9

IUPAC Name: 3-(benzyloxy)pyrazin-2-amine
Synonyms: 3-(benzyloxy)pyrazin-2-amine
Molecular Weight (g/mol): 201.23
Molecular Formula: C11H11N3O
InChi Key: QKEQFJXWHGVJJU-UHFFFAOYSA-N
SMILES: NC1=NC=CN=C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results