accessibility menu, dialog, popup

UserName

Viewing profile as user:
P

CAS RN 110231-30-6

IUPAC Name: diphenyl(pyren-1-yl)phosphane
Synonyms: diphenyl(pyren-1-yl)phosphane
Molecular Weight (g/mol): 386.43
Molecular Formula: C28H19P
InChi Key: DSYGKYCYNVHCNQ-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results