CAS RN 1103535-99-4
IUPAC Name:
3,6-dibromo-9-heptyl-9H-carbazole
Synonyms:
3,6-dibromo-9-heptylcarbazole
Molecular Weight (g/mol):
423.19
Molecular Formula:
C19H21Br2N
InChi Key:
DTRIORAMBSBROF-UHFFFAOYSA-N
SMILES:
CCCCCCCN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2
1
–
1
of
1
results
1
3,6-Dibromo-9-heptyl-9H-carbazole 98.0+%, TCI America™
CAS: 1103535-99-4 Molecular Formula: C19H21Br2N Molecular Weight (g/mol): 423.192 InChI Key: DTRIORAMBSBROF-UHFFFAOYSA-N PubChem CID: 129319316 IUPAC Name: 3,6-dibromo-9-heptylcarbazole SMILES: CCCCCCCN1C2=C(C=C(C=C2)Br)C3=C1C=CC(=C3)Br