CAS RN 110429-36-2
CAS RN 110429-36-2
IUPAC Name:
3-[(2H-1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-1-methylpiperidine
Synonyms:
3-[(2H-1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-1-methylpiperidine
Molecular Weight (g/mol):
343.4
Molecular Formula:
C20H22FNO3
InChi Key:
MOJZPKOBKCXNKG-UHFFFAOYNA-N
SMILES:
CN1CCC(C(COC2=CC3=C(OCO3)C=C2)C1)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
N-Methyl Paroxetine 98.0+%, TCI America™
CAS: 110429-36-2 Molecular Formula: C20H22FNO3 Molecular Weight (g/mol): 343.40 MDL Number: MFCD03788781 InChI Key: MOJZPKOBKCXNKG-UHFFFAOYNA-N PubChem CID: 10947895 IUPAC Name: 3-[(2H-1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-1-methylpiperidine SMILES: CN1CCC(C(COC2=CC3=C(OCO3)C=C2)C1)C1=CC=C(F)C=C1