accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110453-78-6

IUPAC Name: 6-methyloctan-1-ol
Synonyms: 6-methyloctan-1-ol
Molecular Weight (g/mol): 144.26
Molecular Formula: C9H20O
InChi Key: WWRGKAMABZHMCN-UHFFFAOYNA-N
SMILES: CCC(C)CCCCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results