accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1105511-66-7

IUPAC Name: 2-methoxy-6-(tributylstannyl)pyrazine
Synonyms: 2-methoxy-6-(tributylstannyl)pyrazine
Molecular Weight (g/mol): 399.17
Molecular Formula: C17H32N2OSn
InChi Key: WWRXJELRWGUPKZ-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=NC(OC)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results