accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1105665-08-4

IUPAC Name: 5-bromo-2-(morpholin-4-yl)benzonitrile
Synonyms: 5-bromo-2-(morpholin-4-yl)benzonitrile
Molecular Weight (g/mol): 267.13
Molecular Formula: C11H11BrN2O
InChi Key: IENSJQPUDLLAOI-UHFFFAOYSA-N
SMILES: BrC1=CC(C#N)=C(C=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results