accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHNHO(E/Z)

CAS RN 1105698-15-4

IUPAC Name: N-(3-{[(2-oxo-1,2-dihydronaphthalen-1-ylidene)methyl]amino}phenyl)-2-phenylpropanamide
Synonyms: N-(3-{[(2-oxonaphthalen-1-ylidene)methyl]amino}phenyl)-2-phenylpropanamide
Molecular Weight (g/mol): 394.47
Molecular Formula: C26H22N2O2
InChi Key: UADRPWLRVLBVBC-UHFFFAOYNA-N
SMILES: CC(C(=O)NC1=CC=CC(NC=C2C(=O)C=CC3=CC=CC=C23)=C1)C1=CC=CC=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results