CAS RN 11061-68-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Human Insulin Solution, 100 μg/mL in PBS, Sold by MilliporeSigma™ Supelco™
Certified reference material