accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(S)O(S)CH3(R)OH(R)HO(R)O(S)O(S)CH3(R)HO(R)HO(R)OHOOHO(S)O(R)OH(S)OH(S)OH(R)HOH3CCH3O

CAS RN 110642-44-9

IUPAC Name: 3-{[(2S,3R,4R,5R,6S)-4,5-dihydroxy-6-methyl-3-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-2-yl]oxy}-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4H-chromen-4-one
Synonyms: 3-{[(2S,3R,4R,5R,6S)-4,5-dihydroxy-6-methyl-3-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-2-yl]oxy}-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}chromen-4-one
Molecular Weight (g/mol): 822.81
Molecular Formula: C39H50O19
InChi Key: ULZLIYVOYYQJRO-JIYCBSMMSA-N
SMILES: COC1=CC=C(C=C1)C1=C(O[C@@H]2O[C@@H](C)[C@H](O)[C@@H](O)[C@H]2O[C@@H]2O[C@@H](C)[C@H](O)[C@@H](O)[C@H]2O)C(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)C(CC=C(C)C)=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results