CAS RN 11070-44-3
CAS RN 11070-44-3
IUPAC Name:
3a-methyl-1,3,3a,4,5,6-hexahydro-2-benzofuran-1,3-dione
Synonyms:
3a-methyl-5,6-dihydro-4H-2-benzofuran-1,3-dione
Molecular Weight (g/mol):
166.18
Molecular Formula:
C9H10O3
InChi Key:
LOYDTBZMMPQJNI-UHFFFAOYNA-N
SMILES:
CC12CCCC=C1C(=O)OC2=O
1
–
1
of
1
results
1
Methylcyclohexene-1,2-dicarboxylic Anhydride (mixture of isomers) 80.0+%, TCI America™
CAS: 11070-44-3 Molecular Formula: C9H10O3 MDL Number: MFCD00014585 Synonym: Methyltetrahydrophthalic Anhydride