accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110726-28-8

IUPAC Name: 4-[1-(4-hydroxyphenyl)-1-{4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl}ethyl]phenol
Synonyms: 4-[1-(4-hydroxyphenyl)-1-{4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl}ethyl]phenol
Molecular Weight (g/mol): 424.54
Molecular Formula: C29H28O3
InChi Key: WXYSZTISEJBRHW-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=C(O)C=C1)C1=CC=C(C=C1)C(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results