accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOOHNHOO

CAS RN 110797-35-8

IUPAC Name: tert-butyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxypropanoate
Synonyms: tert-butyl 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-hydroxypropanoate
Molecular Weight (g/mol): 383.44
Molecular Formula: C22H25NO5
InChi Key: ZYOWIDHANLLHNO-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C(CO)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results