accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110802-84-1

IUPAC Name: dihydrogen hexachloroiridiumtetrakis(ylium)
Synonyms: dihydrogen hexachloroiridiumtetrakis(ylium)
Molecular Weight (g/mol): 406.93
Molecular Formula: Cl6H2Ir
InChi Key: IBMVGQBDTOACBV-UHFFFAOYSA-J
SMILES: [H+].[H+].Cl[Ir+4](Cl)(Cl)(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results