accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110851-66-6

IUPAC Name: 3-tetradecylthiophene
Synonyms: 3-tetradecylthiophene
Molecular Weight (g/mol): 280.51
Molecular Formula: C18H32S
InChi Key: CAEIOINMYGTXNS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC1=CSC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results