CAS RN 110860-92-9
IUPAC Name:
5,6-dichloropyridin-3-ol
Synonyms:
5,6-dichloropyridin-3-ol
Molecular Weight (g/mol):
163.99
Molecular Formula:
C5H3Cl2NO
InChi Key:
GMHPKIGFTDRQBP-UHFFFAOYSA-N
SMILES:
OC1=CC(Cl)=C(Cl)N=C1
1
–
1
of
1
results
1
2,3-Dichloro-5-hydroxypyridine 98.0+%, TCI America™
CAS: 110860-92-9 Molecular Formula: C5H3Cl2NO Molecular Weight (g/mol): 163.985 MDL Number: MFCD09835219 InChI Key: GMHPKIGFTDRQBP-UHFFFAOYSA-N Synonym: 5,6-Dichloro-3-pyridinol PubChem CID: 14391672 IUPAC Name: 5,6-dichloropyridin-3-ol SMILES: C1=C(C=NC(=C1Cl)Cl)O