accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110876-52-3

IUPAC Name: (2-methylnaphthalen-1-yl)(naphthalen-2-yl)methanone
Synonyms: (2-methylnaphthalen-1-yl)(naphthalen-2-yl)methanone
Molecular Weight (g/mol): 296.37
Molecular Formula: C22H16O
InChi Key: GCDFSVMEIZNGPZ-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=CC2=C1C(=O)C1=CC=C2C=CC=CC2=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results